siRNA Details
| Gene Symbol: RN7SL832P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036490.2 | - | 1325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RN7SL832P| Gene Symbol: RN7SL832P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036490.2 | - | 1325 | |||