siRNA Details
| Gene Symbol: LOC101929076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120345.1 | 2 | 492 | |||
| GenBank mRNA | BC033162.1 | - | 2 | 492 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502156.1 | - | 2 | 492 | Chr. 15 - 67834310 - 67838879 [-] on Build GRCh38 | |

LOC101929076
Genomic Map