siRNA Details
Gene Symbol: LOC100506504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542503.2 | XP_011540805.1 | 2 | 1166 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024190.1 | - | 1 | 1872 | Chr. 1 - 2181794 - 2184053 [-] on Build GRCh38 |
