This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other B230217C12RIK SIRNA Assays ›
Gene Symbol:
B230217C12Rik
Gene Name:
RIKEN cDNA B230217C12 gene
Gene Aliases:
AI840637
Chromosome Location:
Chr.11: 97840780 - 97843043 on Build GRCm38
UniGene ID:
Mm.110270
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n377632

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: B230217C12Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK018341.1 - 2 1324
AK031146.1 - 1323
AK035650.1 - 1306
AK042827.1 - 1326
AK080808.1 - 1323
AK081899.1 - 1344

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-8bmkk:80/100.66.79.215:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0