siRNA Details
| Gene Symbol: 1700024I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006304.1 | - | 5 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700024I08Rik| Gene Symbol: 1700024I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006304.1 | - | 5 | 723 | ||