This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other REST SIRNA Assays ›
Gene Symbol:
Rest
Gene Name:
RE1-silencing transcription factor
Gene Aliases:
2610008J04Rik, AA407358, NRSF, REST4
Chromosome Location:
Chr.5: 77265494 - 77286432 on Build GRCm38
UniGene ID:
Mm.28840
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n379704

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rest
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK011349.1 - 1 1389
BC023433.1 - 1 102
BC051648.1 - 1 438
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CA531486.1 - 1 438 Chr. 5 - 77285367 - 77285900 [+] on Build GRCm38
GenBankmRNA BU961832.1 - 1 260 Chr. 5 - 77285545 - 77286245 [+] on Build GRCm38
GenBankmRNA CN682158.1 - 1 302 Chr. 5 - 77285500 - 77286044 [+] on Build GRCm38
GenBankmRNA CA978936.1 - 1 779 Chr. 5 - 77285025 - 77285909 [+] on Build GRCm38
GenBankmRNA AI663913.1 - 1 466 Chr. 5 - 77285338 - 77285960 [+] on Build GRCm38
GenBankmRNA CF550744.1 - 1 237 Chr. 5 - 77285566 - 77286341 [+] on Build GRCm38
GenBankmRNA CO813766.1 - 325 Chr. - - [-] on Build
GenBankmRNA CB056056.1 - 1 223 Chr. 5 - 77285593 - 77286128 [+] on Build GRCm38
GenBankmRNA BG090150.1 - 1 508 Chr. 5 - 77285296 - 77285834 [+] on Build GRCm38
GenBankmRNA BQ573545.1 - 1 243 Chr. 5 - 77285561 - 77286333 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0