siRNA Details
| Gene Symbol: Gm38575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038448.1 | - | 242 | |||
| AK165297.1 | - | 173 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm38575| Gene Symbol: Gm38575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038448.1 | - | 242 | |||
| AK165297.1 | - | 173 | ||||