siRNA Details
| Gene Symbol: AU015791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102381.1 | 5 | 763 | |||
| GenBank mRNA | AK139027.1 | - | 2 | 1251 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV381755.2 | - | 3 | 464 | Chr. 12 - 105513734 - 105515246 [+] on Build GRCm38 | |

AU015791
Genomic Map