siRNA Details
Gene Symbol: 1700007F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040538.1 | 6 | 719 | |||
GenBank mRNA | AK005707.1 | - | 6 | 700 | ||
AK017083.1 | - | 6 | 721 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700007F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040538.1 | 6 | 719 | |||
GenBank mRNA | AK005707.1 | - | 6 | 700 | ||
AK017083.1 | - | 6 | 721 |