siRNA Details
| Gene Symbol: Gm5606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053768.1 | - | 1 | 1007 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB546007.2 | - | 1 | 223 | Chr. 8 - 12390172 - 12390855 [-] on Build GRCm38 | |

Gm5606
Genomic Map