siRNA Details
| Gene Symbol: 4930504B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015694.1 | - | 1 | 658 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV264942.1 | - | 1 | 170 | Chr. 19 - 14084118 - 14084378 [+] on Build GRCm38 | |

4930504B16Rik
Genomic Map