siRNA Details
| Gene Symbol: AI854517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040311.1 | 5 | 3393 | |||
| NR_040312.1 | 4 | 3275 | ||||
| NR_040313.1 | 3 | 3202 | ||||
| NR_040314.1 | 3 | 3155 | ||||
| GenBank mRNA | AK032343.1 | - | 3 | 3059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI854517| Gene Symbol: AI854517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040311.1 | 5 | 3393 | |||
| NR_040312.1 | 4 | 3275 | ||||
| NR_040313.1 | 3 | 3202 | ||||
| NR_040314.1 | 3 | 3155 | ||||
| GenBank mRNA | AK032343.1 | - | 3 | 3059 | ||