This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM5108 SIRNA Assays ›
Gene Symbol:
Gm5108
Gene Name:
predicted gene 5108
Gene Aliases:
EG330097
Chromosome Location:
Chr.5: 67941669 - 67977070 on Build GRCm38
UniGene ID:
Mm.78496
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n390022

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm5108
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK076825.1 - 1, 2 203
BC120772.1 - 1, 2 131
BC120798.1 - 1, 2 131
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AV264477.2 - 1, 2 203 Chr. 5 - 67941669 - 67977070 [+] on Build GRCm38
GenBankmRNA BB615379.1 - 1, 2 203 Chr. 5 - 67941669 - 67977072 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline