siRNA Details
| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004787.1 | - | 1 | 2129 | ||
| BC037482.1 | - | 1 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndst1| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004787.1 | - | 1 | 2129 | ||
| BC037482.1 | - | 1 | 377 | |||