siRNA Details
| Gene Symbol: Ndrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090374.1 | - | 13, 14 | 964 | ||
| AK144294.1 | - | 13, 14 | 965 | |||
| AK168873.1 | - | 13, 14 | 966 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129305.1 | - | 1277 | Chr. - - [-] on Build | ||

Ndrg4
Genomic Map