siRNA Details
| Gene Symbol: Mym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF019615.2 | - | 1 | 243 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA327347.1 | - | 1 | 525 | Chr. 5 - 28168195 - 28168960 [+] on Build GRCm38 | |

Mym
Genomic Map