siRNA Details
| Gene Symbol: Gm34816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136242.1 | - | 2 | 252 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720324.1 | - | 2 | 252 | Chr. 1 - 139408278 - 139413868 [+] on Build GRCm38 | |

Gm34816
Genomic Map