siRNA Details
| Gene Symbol: 9530078K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020651.1 | - | 1 | 355 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722277.1 | - | 1 | 355 | Chr. 7 - 92902179 - 92902812 [+] on Build GRCm38 | |

9530078K11Rik
Genomic Map