siRNA Details
| Gene Symbol: Pantr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011788.1 | - | 1 | 121 | ||
| AK011913.1 | - | 1 | 121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pantr2| Gene Symbol: Pantr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011788.1 | - | 1 | 121 | ||
| AK011913.1 | - | 1 | 121 | |||