siRNA Details
| Gene Symbol: Gm30459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140552.1 | - | 2 | 278 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ061097.1 | - | 2 | 278 | Chr. 7 - 74765987 - 74766865 [+] on Build GRCm38 | |

Gm30459
Genomic Map