siRNA Details
| Gene Symbol: 9630014C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020679.1 | - | 1 | 516 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722433.1 | - | 1 | 516 | Chr. 2 - 90548419 - 90549085 [-] on Build GRCm38 | |

9630014C11Rik
Genomic Map