siRNA Details
| Gene Symbol: 4930522O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040665.1 | 1 | 79 | |||
| GenBank mRNA | AK019691.1 | - | 1 | 81 | ||
| BY715425.1 | - | 1 | 81 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930522O17Rik| Gene Symbol: 4930522O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040665.1 | 1 | 79 | |||
| GenBank mRNA | AK019691.1 | - | 1 | 81 | ||
| BY715425.1 | - | 1 | 81 | |||