siRNA Details
| Gene Symbol: Gm20757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046027.1 | 5 | 639 | |||
| GenBank mRNA | AK053604.1 | - | 5 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20757| Gene Symbol: Gm20757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046027.1 | 5 | 639 | |||
| GenBank mRNA | AK053604.1 | - | 5 | 640 | ||