siRNA Details
| Gene Symbol: Fem1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031720.1 | - | 2187 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fem1c| Gene Symbol: Fem1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031720.1 | - | 2187 | |||