siRNA Details
| Gene Symbol: 1700047A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110583.1 | 4 | 393 | |||
| GenBank mRNA | AK006702.1 | - | 4 | 394 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706693.1 | - | 4 | 394 | Chr. 8 - 26082609 - 26096511 [+] on Build GRCm38 | |

1700047A11Rik
Genomic Map