siRNA Details
| Gene Symbol: C030010L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021061.1 | - | 2 | 387 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728132.1 | - | 2 | 384 | Chr. 16 - 97994647 - 97997417 [-] on Build GRCm38 | |

C030010L15Rik
Genomic Map