siRNA Details
| Gene Symbol: Gm11419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131774.1 | - | 1 | 252 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707204.1 | - | 1 | 252 | Chr. 11 - 81985406 - 81988769 [+] on Build GRCm38 | |

Gm11419
Genomic Map