siRNA Details
| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040757.1 | 4 | 282 | |||
| GenBank mRNA | AK002866.1 | - | 4 | 283 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703364.1 | - | 4 | 283 | Chr. 6 - 108610333 - 108623167 [-] on Build GRCm38 | |

0610040F04Rik
Genomic Map