siRNA Details
| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040757.1 | 5 | 747 | |||
| GenBank mRNA | AK002866.1 | - | 5 | 748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610040F04Rik| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040757.1 | 5 | 747 | |||
| GenBank mRNA | AK002866.1 | - | 5 | 748 | ||