siRNA Details
| Gene Symbol: Gm11186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046047.1 | 2 | 246 | |||
| GenBank mRNA | AK043416.1 | - | 2 | 246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11186| Gene Symbol: Gm11186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046047.1 | 2 | 246 | |||
| GenBank mRNA | AK043416.1 | - | 2 | 246 | ||