siRNA Details
| Gene Symbol: Fendrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004221.1 | - | 3 | 1026 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB365376.2 | - | 2 | 629 | Chr. 8 - 121058803 - 121059584 [-] on Build GRCm38 | |

Fendrr
Genomic Map