This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IGKV4-53 SIRNA Assays ›
Gene Symbol:
Igkv4-53 Igkv4-90
Gene Name:
immunoglobulin kappa variable 4-53
immunoglobulin kappa chain variable 4-90
Gene Aliases:
Gm1420, Gm1526, IgVkkh4, kh4
Gm16906, an4
Chromosome Location:
Chr.6: 69648824 - 69649354 on Build GRCm38
Chromosome Location:
Chr.6: 68807180 - 68807708 on Build GRCm38
UniGene ID:
Mm.305097
Mm.424510
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID n408866

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Igkv4-53
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC106159.1 - 345
Gene Symbol: Igkv4-90
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA X14098.1 - 287
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG961850.1 - 328 Chr. - - [-] on Build
GenBankmRNA AF137622.1 - 219 Chr. - - [-] on Build
GenBankmRNA DQ372797.1 - 302 Chr. - - [-] on Build
GenBankmRNA AF057545.1 - 245 Chr. - - [-] on Build
GenBankmRNA FR689230.1 - 79 Chr. - - [-] on Build

Back To Top

More Information


Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline