siRNA Details
| Gene Symbol: 9530020I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020565.1 | - | 3 | 523 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722031.1 | - | 3 | 523 | Chr. 12 - 11203086 - 11204409 [-] on Build GRCm38 | |

9530020I12Rik
Genomic Map