This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930453H23RIK SIRNA Assays ›
Gene Symbol:
4930453H23Rik
Gene Name:
RIKEN cDNA 4930453H23 gene
Gene Aliases:
-
Chromosome Location:
Chr.X: 21206580 - 21221672 on Build GRCm38
UniGene ID:
Mm.272069
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n413100

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930453H23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK015451.1 - 4 885
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB274217.1 - 3 480 Chr. X - 21218410 - 21221564 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0