siRNA Details
| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_040709.1 | 4 | 1426 | |||
| GenBank mRNA | AK044422.1 | - | LIT3369 RNCR3 |
4 | 1427 | |
| BC030402.1 | - | 2 | 1211 | |||

A930011O12Rik
Genomic Map