siRNA Details
| Gene Symbol: Slc25a53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012963.1 | - | 7 | 1362 | ||
| AK013345.1 | - | 7 | 1388 | |||
| AK028496.1 | - | 7 | 1354 | |||
| AK049939.1 | - | 2 | 1731 | |||
| BC024604.1 | - | 7 | 1370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a53| Gene Symbol: Slc25a53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012963.1 | - | 7 | 1362 | ||
| AK013345.1 | - | 7 | 1388 | |||
| AK028496.1 | - | 7 | 1354 | |||
| AK049939.1 | - | 2 | 1731 | |||
| BC024604.1 | - | 7 | 1370 | |||