siRNA Details
| Gene Symbol: Gm5602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138132.1 | - | 2 | 386 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY723690.1 | - | 2 | 386 | Chr. 7 - 130862390 - 130866330 [-] on Build GRCm38 | |

Gm5602
Genomic Map