siRNA Details
| Gene Symbol: A730082K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040319.1 | 4 | 439 | |||
| GenBank mRNA | AK136021.1 | - | 4 | 425 | ||
| BG082928.2 | - | 4 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730082K24Rik| Gene Symbol: A730082K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040319.1 | 4 | 439 | |||
| GenBank mRNA | AK136021.1 | - | 4 | 425 | ||
| BG082928.2 | - | 4 | 406 | |||