siRNA Details
| Gene Symbol: 4930550I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016087.1 | - | 1 | 26 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715689.1 | - | 1 | 26 | Chr. 2 - 72629018 - 72630432 [-] on Build GRCm38 | |

4930550I04Rik
Genomic Map