siRNA Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018898.1 | - | 1 | 13 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047531.1 | - | 1 | 29 | Chr. 1 - 130163831 - 130180461 [+] on Build GRCm38 | |

Thsd7b
Genomic Map