siRNA Details
| Gene Symbol: 1700124M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007272.1 | - | 2 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700124M09Rik| Gene Symbol: 1700124M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007272.1 | - | 2 | 199 | ||