siRNA Details
| Gene Symbol: 1700124M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007272.1 | - | 2 | 112 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707371.1 | - | 2 | 112 | Chr. 10 - 19086527 - 19087178 [-] on Build GRCm38 | |

1700124M09Rik
Genomic Map