siRNA Details
| Gene Symbol: 1700016P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038149.1 | 2 | 410 | |||
| GenBank mRNA | AK006052.1 | - | 2 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700016P04Rik| Gene Symbol: 1700016P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038149.1 | 2 | 410 | |||
| GenBank mRNA | AK006052.1 | - | 2 | 413 | ||