siRNA Details
| Gene Symbol: Gm27198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133101.1 | - | 2 | 108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615927.1 | - | 2 | 108 | Chr. 14 - 118728650 - 118735145 [-] on Build GRCm38 | |

Gm27198
Genomic Map