siRNA Details
| Gene Symbol: 4933417C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016840.1 | - | 1 | 62 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716515.1 | - | 1 | 62 | Chr. 1 - 160213694 - 160214505 [+] on Build GRCm38 | |

4933417C20Rik
Genomic Map