siRNA Details
| Gene Symbol: Macrod2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031201.1 | - | 4 | 954 | ||
| Gene Symbol: 4931432E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016500.1 | - | 1 | 828 | ||

Macrod2os1
Genomic Map