siRNA Details
| Gene Symbol: Gm4719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045952.1 | 4 | 467 | |||
| GenBank mRNA | AK131717.1 | - | 4 | 468 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706359.1 | - | 4 | 463 | Chr. 17 - 89379245 - 89384944 [+] on Build GRCm38 | |

Gm4719
Genomic Map