siRNA Details
| Gene Symbol: Gm32693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039352.1 | - | 3 | 465 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB633099.1 | - | 3 | 462 | Chr. 3 - 139778859 - 139780758 [-] on Build GRCm38 | |

Gm32693
Genomic Map