siRNA Details
| Gene Symbol: 2900052O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013671.1 | - | 1 | 685 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712650.1 | - | 1 | 685 | Chr. 10 - 32097535 - 32098349 [-] on Build GRCm38 | |

2900052O17Rik
Genomic Map