siRNA Details
| Gene Symbol: 1700085D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007005.1 | - | 2 | 99 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707061.1 | - | 2 | 95 | Chr. 15 - 73249546 - 73264120 [+] on Build GRCm38 | |

1700085D07Rik
Genomic Map